Anti-EFNA5 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP65431_P050
AVIVARP65431_P050EA 401 EUR
AVIVARP65431_P050
Anti-EFNA5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EFNA5 (ephrin-A5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human