Anti-KIR2DL5A Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP65773_P050
AVIVARP65773_P050EA 401 EUR
AVIVARP65773_P050
Anti-KIR2DL5A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KIR2DL5A (killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human