Anti-MRPL12 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP65789_P050
AVIVARP65789_P050EA 401 EUR
AVIVARP65789_P050
Anti-MRPL12 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MRPL12 (mitochondrial ribosomal protein L12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR