Anti-RPTN Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP65825_P050
AVIVARP65825_P050EA 401 EUR
AVIVARP65825_P050
Anti-RPTN Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RPTN (repetin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human