Anti-VCY Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP65848_P050
AVIVARP65848_P050EA 401 EUR
AVIVARP65848_P050
Anti-VCY Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: VCY (variable charge, Y-linked)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR