Anti-TNNC2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP65981_P050
AVIVARP65981_P050EA 401 EUR
AVIVARP65981_P050
Anti-TNNC2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TNNC2 (troponin C type 2 (fast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen