Anti-FLVCR2 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP66352_P050
AVIVARP66352_P050EA 401 EUR
AVIVARP66352_P050
Anti-FLVCR2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FLVCR2 (feline leukemia virus subgroup C cellular receptor family, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

TECHNISCHE DATEN

Antigen-Symbol FLVCR2
Environmentally Preferable
Antigen-Name Feline leukemia virus subgroup C cellular receptor 2
Antigen-Synonyme PVHH|C14orf58|MFSD7C|CCT|FLVCRL14q|EPV
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human
Wirt Rabbit
Gen-ID 55640
Isotyp
Western blot Yes
Epitop C-terminal
Kreuzadsorption none
Lagertemperatur Cool
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR