Anti-IDH3B Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP66591_P050
AVIVARP66591_P050EA 401 EUR
AVIVARP66591_P050
Anti-IDH3B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IDH3B (isocitrate dehydrogenase 3 (NAD+) beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR