Anti-HAUS3 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP66821_P050
AVIVARP66821_P050EA 401 EUR
AVIVARP66821_P050
Anti-HAUS3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HAUS3 (HAUS augmin-like complex, subunit 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human