Anti-IYD Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP66837_P050
AVIVARP66837_P050EA 401 EUR
AVIVARP66837_P050
Anti-IYD Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IYD (iodotyrosine deiodinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human