Anti-6330545A04Rik Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP67275_P050
AVIVARP67275_P050EA 401 EUR
AVIVARP67275_P050
Anti-6330545A04Rik Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: 6330545A04Rik
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR