Anti-H2AFV Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP67546_P050
AVIVARP67546_P050EA 401 EUR
AVIVARP67546_P050
Anti-H2AFV Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: H2AFV (H2A histone family, member V)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human