Anti-DHX40 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP68278_P050
AVIVARP68278_P050EA 401 EUR
AVIVARP68278_P050
Anti-DHX40 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DHX40 (DEAH (Asp-Glu-Ala-His) box polypeptide 40)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR