Anti-EPS15L1 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP68294_P050
AVIVARP68294_P050EA 401 EUR
AVIVARP68294_P050
Anti-EPS15L1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EPS15L1 (epidermal growth factor receptor pathway substrate 15-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR