Anti-KATNBL1 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP68900_P050
AVIVARP68900_P050EA 401 EUR
AVIVARP68900_P050
Anti-KATNBL1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KATNBL1 (katanin p80 subunit B-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR