Anti-C16orf59 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP68949_P050
AVIVARP68949_P050EA 401 EUR
AVIVARP68949_P050
Anti-C16orf59 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: C16orf59 (chromosome 16 open reading frame 59)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR