Anti-ZMYND15 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP69005_P050
AVIVARP69005_P050EA 401 EUR
AVIVARP69005_P050
Anti-ZMYND15 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZMYND15 (zinc finger, MYND-type containing 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR