Anti-SKIDA1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP69749_P050
AVIVARP69749_P050EA 401 EUR
AVIVARP69749_P050
Anti-SKIDA1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SKIDA1 (SKI/DACH domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human