Anti-FIGNL2 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP70421_P050
AVIVARP70421_P050EA 401 EUR
AVIVARP70421_P050
Anti-FIGNL2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FIGNL2 (fidgetin-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR