Anti-FRMD1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP70435_P050
AVIVARP70435_P050EA 401 EUR
AVIVARP70435_P050
Anti-FRMD1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FRMD1 (FERM domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human