Anti-PRSS45 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP71468_P050
AVIVARP71468_P050EA 401 EUR
AVIVARP71468_P050
Anti-PRSS45 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRSS45 (protease, serine, 45)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human