Anti-CDK1 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVARP03024P050
AVIVAVARP03024P050EA 401 EUR
AVIVAVARP03024P050
Anti-CDK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR