1H,1H,2H-Perfluor-1-octen ≥99%

Lieferant: Thermo Fisher Scientific

B20920.06 B20920.14
B20920.06EA 40.3 EUR
B20920.06 B20920.14
1H,1H,2H-Perfluor-1-octen ≥99%
1H,1H,2H-Perfluor-1-octen

Purity: 99% CAS No.: 25291-17-2 Molecular Formula: CF3(CF2)5CH=CH2 Formula Weight: 346.09 Flash Point: 20°(202°F) Refractive Index: 1.2960 Boiling Point: 106-107° Beilstein Reference: 2060994 MDL No.: MFCD00039249

Formel: C₈H₃F₁₃
Siedepunkt: 106…107 °C
Dichte: 1,580
Flammpunkt: 20 °C (202 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00039249
CAS-Nummer: 25291-17-2
EINECS: 246-791-8
UN: 1993
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR