Experimentation kits, antigen-antibody interactions

Lieferant: G-Biosciences
BE-501
BE-501EA 374 EUR
BE-501
Experimentation kits, antigen-antibody interactions
Lehrmittel Biologie-Lehrmittel Physiology and Health Learning Activities
This lab activity is designed to study highly specific lock-key matching properties of antigen-antibody and how this highly specific interaction can be exploited as a tool for research and analysis. This study involves the use of an immunodiffusion technique in which antigen and antibody are allowed to diffuse in a solid agarose medium. When antigen and antibody meet, antigen-antibody complex is formed, which leads to precipitation. Antigen-antibody precipitate is formed in the zone where the concentration of the two matching pairs reaches an optimal known as the zone of equivalence, which results in formation of a visible opaque precipitate region in agarose medium. Those regions of precipitation can be used for determination of concentration or titer of both antigen and antibody. This Antigen-Antibody Interaction kit is a hands-on study of both Ouchterlony Double Diffusion and Radial Immunodiffusion techniques. This kit also provides additional guidance materials for teaching other types of antigen-antibody interactions concepts such as immunoelectrophoresis and immunoprecipitation.

  • Specific properties of antigen and antibody
  • Antigen-antibody diffusion, interaction, and complex formation
  • Teaches Ouchterlony Double Diffusion and Radial Immunodiffusion
  • Application of antigen-antibody interaction in research laboratories

Also required: Water baths or beakers and thermometer.

Lieferumfang: Supplied with components needed for hands-on experimentation for six workstations of 4 to 5 students or 24 to 30 students. Supplied with Teacher’s Guide and separate Student’s Guides.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR