Human skull model
Lieferant: 3B Scientific
1000049
BINH1000049EA 0 EUR
BINH1000049
Human skull model
Modelle Humanbiologie
The three-part medical quality human skull replica is designed for basic anatomical studies of the skull. It includes an anatomically correct five-part brain.
  • High quality original skull cast
  • Handmade from hard plastic skull
  • Highly accurate representation of the fissures, foramina, processes, sutures
  • Can be disassembled into skull cap, base of skull, and mandible
  • Mandible of skull is mounted on a spring to easily demonstrate natural movement
This medical grade skull model also makes a great present for medical professionals and students of medicine and allied health professions.