Transparent classic human skull model
Lieferant: 3B Scientific
1000051
BINH1000051EA 0 EUR
BINH1000051
Transparent classic human skull model
Modelle Humanbiologie
Use this unique transparent replica of the human skull to study internal structures that otherwise are visible only through x-ray images.
  • High-quality original casts
  • Skull is handmade of hard, unbreakable plastic
  • Highly accurate representation of the fissures, foramina, processes, sutures etc.
  • Can be disassembled into Skull Cap, Base of Skull and Mandible
  • As an option, you can insert a five-part brain (C 18) into the skull