Human skull model, demonstrates functions of masticator muscles, 2 part
Lieferant: 3B Scientific
1000056
BINH1000056EA 607 EUR
BINH1000056
Human skull model, demonstrates functions of masticator muscles, 2 part
Modelle Humanbiologie
In this human skull model the masticatory muscles (masseter, temporal, medial and lateral pterygoid muscles) are represented as elastic bands. This skull model is suited to demonstrate the function of the masticator muscles with jaw occlusion, the initial stage of jaw opening and the movements of the mandible to the side and front.
  • The skullcap is removable
  • High-quality original casts
  • Skull is handmade of hard, unbreakable plastic
  • Highly accurate representation of the fissures, foramina, processes, sutures etc.
Weight: 0,7 kg