Cervical spinal column
Lieferant: 3B Scientific
1000144
BINH1000144EA 195 EUR
BINH1000144
Cervical spinal column
Modelle Humanbiologie
This real life replica of the anatomy of the cervical spinal column is a high quality model.
  • Consisting of:
  • Occipital plate
  • Seven cervical vertebrae with intervertebral discs
  • Cervical nerves
  • Vertebral arteries
  • Spinal cord
On a flexible stand.