Mini knee joint with cross section
Lieferant: 3B Scientific
1000170
BINH1000170EA 92.3 EUR
BINH1000170
Mini knee joint with cross section
Modelle Humanbiologie
The mini knee joint has been reduced to a half of the natural size of a human knee but has kept all of the functionality of the larger knee joint models. In addition to the external anatomical structures of the knee, the medical or teaching professional now has the possibility to explain what is happening from 'within' the knee joint.
  • Mounted on the base
Dimensions: 10×14×24 cm
Weight: 0,35 kg