Embryo and stages of fertilisation
Lieferant: 3B Scientific
1000320
BINH1000320EA 607 EUR
BINH1000320
Embryo and stages of fertilisation
Modelle Humanbiologie
The model illustrates schematically how the ovum matures, ovulation and fertilisation occurs.
  • Models of ovary, fallopian tube, and womb
  • Enlarged illustrations also printed on the base
It shows how the ovum develops to the stage where it embeds itself in the womb wall to begin the growth into an embryo.
Lieferumfang: Supplied on a base.