Dove wing model
Lieferant: Binhold, Paul Lehrmittel
1002557
BINH1002557EA 0 EUR
BINH1002557
Dove wing model
Modelle Zoologie
Columba palumbus.
  • Presented in a showcase
  • Labelled in English
  • Genuine specimen from an animal skeleton
The wing and feathers are presented for a comparative anatomy, ornithology, and other studies.