Functional ear model
Lieferant: 3B Scientific
1005052
BINH1005052EA 0 EUR
BINH1005052
Functional ear model
Modelle Humanbiologie
This functional ear model shows how the tympanic membrane, ossicles, the complex internal ear with the cochlea and the oscillations of the basilar hearing membrane operate/interact.
  • Enclosed mirror enables operation of the functional ear model
  • One single ear model may be studied by several students simultaneously in an action-oriented learning situation
Suitable for the studying of various ear-functions from different angles at the same time.
Bestellinformation: Functional ear includes a four-colour explanatory chart.