Small molecular model: DNA/RNA
Lieferant: Binhold, Paul Lehrmittel
1005302
BINH1005302EA 63.4 EUR
BINH1005302
Small molecular model: DNA/RNA
Modelle Molekularbiologie
The small DNA/RNA model shows the double helix and how the molecules split at the centre of the base pairs. It includes:
  • 30 coloured tubes – Guanine red, cytosine green, thymine blue, adenine gray and uracil white
  • 12 white two prong centre representing the hydrogen bond between the base pairs
  • 28 black trigonal atom centres showing sugar
  • 25 red two prong atom centres, which represent phosphate groups
  • 50 yellow tubes that link the phosphate groups to the sugar rings
Bestellinformation: Includes a wooden base with support rod.