Venous access device model
Lieferant: 3B Scientific
1005563
BINH1005563EA 2110 EUR
BINH1005563
Venous access device model
Modelle Humanbiologie
Teach patients and healthcare staff the care and use of two different types of catheters with this model. Features left and right subclavian catheter connections that may be flushed with water; implanted venous access system with a 'skin' flap over the implant to allow viewing.
  • Mounted on a collapsible easel
  • Made of skinned and lightweight urethane foam
  • May be washed with soap, water as well as antiseptics
Verpackung: In its carrying case.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable