Anti-CD3G Rat Monoclonal Antibody (Brilliant Violet® 711) [clone: 17A2]

Lieferant: BIOLEGEND INC
100241
BLEG100241EA 433 EUR
BLEG100241
Anti-CD3G Rat Monoclonal Antibody (Brilliant Violet® 711) [clone: 17A2]
Antikörper
Type: Primary
Antigen: CD3G
Clonality: Monoclonal
Clone: 17A2
Conjugation: Brilliant Violet® 711
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR