Anti-CD4 Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: RM4-5]

Lieferant: BIOLEGEND INC
100547
BLEG100547EA 326 EUR
BLEG100547
Anti-CD4 Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: RM4-5]
Antikörper
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RM4-5
Conjugation: Brilliant Violet® 605
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR