Anti-CD8A Rat Monoclonal Antibody (Brilliant Violet® 711) [clone: 53-6.7]

Lieferant: BIOLEGEND INC
100747
BLEG100747EA 326 EUR
BLEG100747
Anti-CD8A Rat Monoclonal Antibody (Brilliant Violet® 711) [clone: 53-6.7]
Antikörper
Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: Brilliant Violet® 711
Epitope: α chain
Host: Rat
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR