Anti-ITGAM Rat Monoclonal Antibody (Brilliant Violet® 711) [clone: M1/70]

Lieferant: BIOLEGEND INC
101241
BLEG101241EA 326 EUR
BLEG101241
Anti-ITGAM Rat Monoclonal Antibody (Brilliant Violet® 711) [clone: M1/70]
Antikörper
Type: Primary
Antigen: ITGAM
Clonality: Monoclonal
Clone: M1/70
Conjugation: Brilliant Violet® 711
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse, Human, Cynomolgus, Rhesus, Baboon, Chimpanzee,Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR