Anti-CD11B Antibody (Brilliant Violet 785) [clone: M1/70]

Lieferant: BIOLEGEND INC
101243
BLEG101243EA 435 EUR
BLEG101243
Anti-CD11B Antibody (Brilliant Violet 785) [clone: M1/70]
Antikörper
Type:
Antigen: CD11B
Clonality:
Clone: M1/70
Conjugation: Brilliant Violet 785
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR