Anti-CD16 + CD32 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: 93]

Lieferant: BIOLEGEND INC
101331
BLEG101331EA 288 EUR
BLEG101331
Anti-CD16 + CD32 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: 93]
Antikörper
Type: Primary
Antigen: CD16 + CD32
Clonality: Monoclonal
Clone: 93
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR