Anti-CD44 Rat Monoclonal Antibody (Brilliant Violet® 570) [clone: IM7]

Lieferant: BIOLEGEND INC
103037
BLEG103037EA 320 EUR
BLEG103037
Anti-CD44 Rat Monoclonal Antibody (Brilliant Violet® 570) [clone: IM7]
Antikörper
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 0
Conjugation: Brilliant Violet® 570
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR