Anti-PTPRC Rat Monoclonal Antibody (Brilliant Violet® 570) [clone: RA3-6B2]

Lieferant: BIOLEGEND INC
103237
BLEG103237EA 312 EUR
BLEG103237
Anti-PTPRC Rat Monoclonal Antibody (Brilliant Violet® 570) [clone: RA3-6B2]
Antikörper
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 570
Epitope:
Host: Rat
Isotype:
Reactivity: Cat, Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR