Anti-ITGA2 Hamster Monoclonal Antibody (Unconjugated) [clone: ]

Lieferant: BIOLEGEND INC
103501
BLEG103501EA 77 EUR
BLEG103501
Anti-ITGA2 Hamster Monoclonal Antibody (Unconjugated) [clone: ]
Antikörper
Type: Primary
Antigen: ITGA2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR