Anti-CD69 Hamster Monoclonal Antibody (Brilliant Violet® 605) [clone: H1.2F3]

Lieferant: BIOLEGEND INC
104529
BLEG104529EA 338 EUR
BLEG104529
Anti-CD69 Hamster Monoclonal Antibody (Brilliant Violet® 605) [clone: H1.2F3]
Antikörper
Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: H1.2F3
Conjugation: Brilliant Violet® 605
Epitope:
Host: Hamster
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR