Anti-CD80 Hamster Monoclonal Antibody (Brilliant Violet® 650) [clone: 16-10A1]

Lieferant: BIOLEGEND INC
104731
BLEG104731EA 370 EUR
BLEG104731
Anti-CD80 Hamster Monoclonal Antibody (Brilliant Violet® 650) [clone: 16-10A1]
Antikörper
Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 16-10A1
Conjugation: Brilliant Violet® 650
Epitope:
Host: Hamster
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR