Anti-CD86 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: ]

Lieferant: BIOLEGEND INC
105031
BLEG105031EA 288 EUR
BLEG105031
Anti-CD86 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: ]
Antikörper
Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: 0
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR