Anti-CD86 Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: GL-1]

Lieferant: BIOLEGEND INC
105037
BLEG105037EA 355 EUR
BLEG105037
Anti-CD86 Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: GL-1]
Antikörper
Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: GL-1
Conjugation: Brilliant Violet® 605
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR