Anti-THY1 Rat Monoclonal Antibody (Unconjugated) [clone: ]

Lieferant: BIOLEGEND INC
105201 105202
BLEG105201EA 77.2 EUR
BLEG105201 BLEG105202
Anti-THY1 Rat Monoclonal Antibody (Unconjugated) [clone: ]
Antikörper
Type: Primary
Antigen: THY1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2c kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR