Anti-ICAM2 Rat Monoclonal Antibody (Unconjugated) [clone: ]

Lieferant: BIOLEGEND INC
105601 105602
BLEG105601EA 113 EUR
BLEG105601 BLEG105602
Anti-ICAM2 Rat Monoclonal Antibody (Unconjugated) [clone: ]
Antikörper
Type: Primary
Antigen: ICAM2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR