Anti-CTLA4 Hamster Monoclonal Antibody (Biotin) [clone: ]

Lieferant: BIOLEGEND INC
106303 106304
BLEG106303EA 102 EUR
BLEG106303 BLEG106304
Anti-CTLA4 Hamster Monoclonal Antibody (Biotin) [clone: ]
Antikörper
Type: Primary
Antigen: CTLA4
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Hamster
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR